Learn More
Applied Biosystems Veriti 96-Well Thermal Cycler
The Applied Biosystems Veriti 96-Well Thermal Cycler delivers the proven reliability of Applied Biosystems instruments.
Manufacturer: Applied Biosystems 4375786
Description
The Applied Biosystems Veriti 96-Well Thermal Cycler delivers the proven reliability of Applied Biosystems instruments.
With the added control of VeriFlex technology, you have six independent temperature blocks that provide precise control over your PCR optimization. Additionally, the color touch screen simplifies setup and use of the Veriti 96-Well Fast Thermal Cycler. Optional setups for fast or standard PCR methods provide with flexibility to shorten your PCR cycling times. The Veriti 96-Well Thermal Cycler is equipped with a standard 0.2mL block configuration.Key Highlights:
- Innovative VeriFlex Blocks allow for precise PCR optimization and enable you to run up to 6 independent assays
- Standard 0.2 mL block format is compatible with many downstream applications
- Standard and fast enabled cycling to address your current and future PCR needs
- Easy-to-use graphical interface (6.5 inch VGA Touch Screen)
- Convenient protocol transfer from one Veriti Thermal Cycler to another using USB
- Reduced PCR cycling time when using faster ramp rates
- Optional VeritiLink Remote Management Software provides you with peace of mind, allowing you to remotely manage more than 50 cyclers from your PC or internet-compatible phone
The Veriti 96-Well Thermal Cycler features innovative VeriFlex Blocks for enhanced PCR functionality. Six separate peltier blocks provide maximal versatility and flexibility and offer two key benefits: 1) PCR Optimization: each block can be set with a specific temperature which is ideal for precise control over PCR optimization 2) Run more experiments: the six peltier blocks can also be used to run up to six different annealing temperatures in the same run. With this enhanced functionality, the Veriti 96-Well Thermal Cycler with VeriFlex blocks is the ideal thermal cycler for any lab.
Control at your fingertips
The powerful, yet simple to operate user interface on the Veriti system is driven by the 6.5 inch (16.51 cm) VGA color touch screen. The large screen allows for easy viewing of your temperature profiles. Additionally, the large navigation buttons put programming of the Veriti Thermal Cycler at your finger tips. Setup and navigation of the Veriti Thermal Cycler does not require the use of a stylus or mouse.
Fast setup, fast results
The Veriti system has two different options for method navigation. For quick setup, you may select one of the pre-programmed methods to run. If you prefer to program your own methods, simply touch the step you would like to setup and then use the keypad to enter thermal cycling steps. After completing a setup, you have the choice of saving the method to the instrument or to a USB memory stick.
Control your time: Go fast when you want
With the Veriti Thermal Cycler you get two thermal cyclers in one, a standard thermal cycler with its better than gradient feature, and a fast enabled thermal cycler. So whether you have a current need to run fast PCR and shorten your PCR cycling times, or a possible future application need for fast PCR, the Veriti Thermal Cycler provides you with the flexibility to go fast when you want.
For Research Use Only. Not for use in diagnostic procedures.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Store at room temperature. | |
6.5 in. VGA 32k color with touch screen | |
3.9°C⁄sec | |
Auto re-start (after power outages), Program overwrite protection | |
10-100 µl | |
± 3.35 °C⁄sec | |
Menu driven through touch screen | |
96 x 0.2 ml tubes, 1 x 96-well plate | |
±0.25°C (35°C to 99.9°C) | |
High-throughput Compatible |
Height: 24.5cm (9.6 in.), Width: 23.7 cm (9.3 in.), Depth: 48.5 cm (19.1 in.) | |
Veriti Thermal Cycler | |
Veriti™ | |
Fast, Standard | |
VeriFlex™ Blocks: 25 °C (5 °C Zone-to-Zone), Standard 0.2 ml format and sample block enabled to run fast chemistry | |
4.0-99.9 °C | |
11.4 kg (25 lb) | |
0.2 ml tubes, 96-well plate | |
<0.5 °C (20 sec after reaching 95 °C) | |
USB and On-board |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.