Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Applied Biosystems™ Veriti™ Thermal Cycler Extended Warranty Package, 96-well Non-distribution product as customer accommodation.
Verit™ Thermal Cyclers deliver the proven reliability of Applied Biosystems™ instruments with the volume, throughput, block format, and pre-programmed method options to accommodate your research needs. This package includes the Verti Thermal Cycler and an extended warranty.
Specifications
For Use With (Equipment) Veriti™ Thermal Cycler
Product Line Veriti™
Warranty Priority access to Applications and Instrument Technical Support teams, Factory-certified refurbished replacement instrument, Parts labor and shipping, Priority access to off-site Remote Service Engineer
Warranty Response Time Replacement instrument shipped in 1 business day and received within 3 days
Warranty Services Not Included User-replaceable parts, Planned maintenance
View More Specs
Products 2
Catalog Number Mfr. No. Duration Price Quantity & Availability  
Catalog Number Mfr. No. Duration Price Quantity & Availability  
A26658 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
A26658/PKG
1 Year REX Extended Warranty
Each for $8,744.00
N/A
A26660 Non-distribution product as customer accommodation.
View Documents
Applied Biosystems™
A26660/PKG
3 Year ABRC Extended Warranty
Each for $10,295.00
N/A
Description
Description
By combining an extended warranty with your instrument purchase, you’ll extend the life of your investment and receive the support you need to keep your instrument running at peak performance. Each Veriti Thermal Cycler Extended Warranty Package includes instrument features and extended warranty benefits to meet your research goals:
  • Standard block formats compatible with many downstream applications
  • Standard and fast enabled cycling to address your current and future PCR needs
  • Easy-to-use graphical interface (6.5 inch VGA Touch Screen) speeds setup time and use
  • Convenient protocol transfer from one Veriti Thermal Cycler to another using USB port
  • Fast repair from factory-trained and certified service engineers
  • Troubleshooting with experienced Technical Support teams
  • Minimal instrument downtime
Rapid Exchange (REX) Extended Warranty
Add additional warranty coverage to your existing manufacturer’s limited warranty with the Rapid Exchange (REX) extended warranty. If your instrument needs repair, just call our Support Center and we’ll send you a factory-certified refurbished replacement instrument within one business day from the time we determine a replacement is required. Instruments are typically received within 3 days of shipment and are yours to keep. All parts, labor, and shipping charges are included, and existing warranty contracts will be transferred to replacement units.
The Rapid Exchange extended warranty is ideal for customers with user-installable instruments that do not require asset tag consistency, and for those operating under critical timelines.
Certain exclusions apply. Contact your Service and Support Representative for details.
Specifications
Specifications
Veriti™ Thermal Cycler
Priority access to Applications and Instrument Technical Support teams, Factory-certified refurbished replacement instrument, Parts labor and shipping, Priority access to off-site Remote Service Engineer
User-replaceable parts, Planned maintenance
Veriti™
Replacement instrument shipped in 1 business day and received within 3 days