Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier KidWind Basic to Advanced Wind Experiment Kit Upgrade Products for Science Education

Upgrade from KidWind Basic Wind Experiment Kit to KidWind Advanced Wind Experiment Kit.

Manufacturer:  Vernier KWBUPA

Catalog No. S16179ND


Add to cart

Description

Description

Kit Components:
  • High torque generator with wires
  • Motor mount pack
  • Graduated cylinder (250mL)
  • Small water pump
  • Clear tubing 304 x 6.35mm (12 x 0.25 in.)
  • 16-tooth gear
  • 64-tooth gear
Specifications

Specifications

Education
Engineering Education, Renewable Energy
KidWind Basic to Advanced Wind Experiment Kit Upgrade
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit