Learn More
Vernier Vernier Spectrum Tube Carousel Power Supply Products for Science Education
Ultra safe carousel design allows seamless switch from one gas to another without handling the tubes.
Manufacturer:  Vernier STCAR
Catalog No. S16257ND
Add to cart
Description
Description
  • Allows viewing of spectral lines using any Venier spectrometer
  • Holds eight gas spectrum tubes
  • Ultrasafe
  • No exposed high voltage
  • Also includes Spectrum Tube Carousel Fiber Optic Holder
  • Gas spectrum tubes sold separately
  • For Grades 9 and up
Specifications
Specifications
Spectrum Tube Carousel Power Supply, Spectrum Tube Carousel Fiber Optic Holder (Has optical fiber in position)
Spectrum Tube Carousel Power Supply