Learn More
Thermo Scientific™ VersaTREK™ Myco Susceptibility Kit
Perform comprehensive Mtb susceptibility testing with a kit that includes three FDA-cleared antituberculosis drugs: rifampin, isoniazid and ethambutol.
Manufacturer:  Thermo Scientific™ Y711560
Catalog No. Y711560
Add to cart
Description
Description
Perform comprehensive Mtb susceptibility testing with three FDA-cleared antituberculosis drugs using the Thermo Scientific™ VersaTREK™ Myco Susceptibility Kit. The kit includes rifampin, isoniazid and ethambutol.
The VersaTREK System combines a liquid culture medium (VersaTREK Myco media), a growth supplement (Myco GS), and, for potentially contaminated species, an antibiotic supplement (Myco AS or Myco PVNA) with a detection system that automatically incubates and continuously monitors culture bottles inoculated with specimens possibly containing mycobacteria.
The VersaTREK Myco Susceptibility Kit consists of three primary antituberculosis drugs: Rifampin (1µg/mL), Isoniazid (0.4µg/mL and 0.1µg/mL), and Ethambutol (8µg/mL and 5µg/mL).
Mycobacteria susceptibility testing also requires VersaTREK Myco Growth Supplement (Myco GS, Part No. 7112-42) and VersaTREK Myco Antibiotic Supplement (Myco AS, Part No. 7114-42).
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
VersaTREK Myco Susceptibility Kit
Mycobacterium tuberculosis
2 x 25 mL Rifampin, 2 x 25 mL Isoniazid, 3 x 25 mL Ethambutol
Rifampin, Isoniazid, and Ethambutol
80 Tests/Kit
VersaTREK Myco Susceptibility Kit
Mycobacteria
80 Tests/Kit
All Samples (e.g. Sputum, Blood, Tissue, Urine, etc.)
M. tuberculosis Susceptibility Testing