Learn More
Thermo Scientific™ Versette™ Automated Liquid Handler
Support a wide range of applications with this compact handler, featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads.
Manufacturer:  Thermo Scientific™ 65002NTC
 View more versions of this product
Catalog No. 14-245-304
  Check Availability
Add to cart
Specifications
Description
Support a wide range of applications with the Thermo Scientific™ Versette™ Automated Liquid Handler, a compact handler featuring user-friendly programming, reliable performance and a choice of 96- or 384-channel pipetting heads. An ideal choice for the liquid handling needs of most laboratories, it supports 96/384 plate replication, plate stamping and serial dilution procedures.
Support a wide range of applications, including high-throughput Mass Spectrometric Immunoassay (MSIA), plate stamping, plate reformatting, serial dilution and many other applications.
96-and 384-channel pipetting heads:
  • Total volume range 0.5-300µL
  • All pipetting heads include RFID tags for self-identification
Consumables for optimized performance:
  • All pipetting heads utilize Thermo Scientific™ D.A.R.T.'S
Compact size:
  • Six position stage uses a unique dual-level design to minimize space requirements
  • Fits on standard bench and biological hoods
User-friendly programming options:
  • Simple pipetting procedures can be created using visual on-board graphical display
  • For more complex protocols and advanced editing, Thermo Scientific™ ControlMate™ software is available
Description & Specifications
Specifications
0.5 to 300µL
18.1kg
40 lb.
96- and 384- channel pipetting module for use with 96- and 384-channel Versette pipetting heads