Learn More
Thermo Scientific™ Versette™ Pipetting Heads
Choose from a volume range of 0.5µL to 300µL with Thermo Scientific™ Versette™ Pipetting Heads.
Manufacturer:  Thermo Scientific™ 650069630
 View more versions of this product
Catalog No. 14-245-500
Add to cart
Description
Description
Compatible with multiple interchangeable pipetting heads, the Thermo Scientific™ Versette™ Automated Liquid Handler provides versatile pipetting options for a broad range of applications that require 96– to 384–channel automated pipetting. The Versette system is compatible with disposable D.A.R.T.’S™, with a total volume ranging from 0.5–300µl.
 The 96– and 384–channel pipetting heads use Thermo Scientific D.A.R.T.’S™, which ensure effective sealing that provides accurate and precise pipetting across all channels.
  • Range from 96- to 384-channels
Specifications
Specifications
2.0% or 0.15µL
96 Channels
1.5% or 0.1 µL
96
0.5 to 30 μL