Thermo Scientific™ Versette™ Pipetting Heads Available on GSA/VA Contract for Federal Government customers only.
Choose from a volume range of 0.5µL to 300µL with Thermo Scientific™ Versette™ Pipetting Heads.
Manufacturer: Thermo Scientific™ 6500696300
 View more versions of this product
Catalog No. 14-245-314
  Check Availability
Add to Cart
    Specifications
    Volume (Metric) Range 5 to 300µL
    Channels 96
    Description 96 channels
    Accuracy 2.0% or 1.0µL
    Precision 1.5% or 0.75µL
    Choose from a volume range of 0.5µL to 300µL with Thermo Scientific™ Versette™ Pipetting Heads. These disposal beads include RFID tags to self-identify.
    • Range from 96- to 384-channels
  • Description & Specifications
Related Products
Product Content Feedback