Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Versette™ Pipetting Heads
Choose from a volume range of 0.5µL to 300µL with Thermo Scientific™ Versette™ Pipetting Heads.
Products 2
Catalog Number Mfr. No. Channels Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Channels Volume (Metric) Price Quantity & Availability  
14-245-500
View Documents
Thermo Scientific™
650069630
96 0.5 to 30 μL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
14-245-501
View Documents
Thermo Scientific™
65006384100
384 1 to 100 μL This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Compatible with multiple interchangeable pipetting heads, the Thermo Scientific™ Versette™ Automated Liquid Handler provides versatile pipetting options for a broad range of applications that require 96– to 384–channel automated pipetting. The Versette system is compatible with disposable D.A.R.T.’S™, with a total volume ranging from 0.5–300µl.
 The 96– and 384–channel pipetting heads use Thermo Scientific D.A.R.T.’S™, which ensure effective sealing that provides accurate and precise pipetting across all channels.
  • Range from 96- to 384-channels
  • All pipetting heads utilize RFID technology for self-identification.