Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ VersiCap Mat, 96-well, domed cap strips

Flexible mats of flat, domed cap strips for PCR or ultra clear cap strips for qPCR applications for use with Thermo Scientific 96-well PCR plates and strip tubes.

$91.00 - $293.00

Specifications

Color Clear
Material Polypropylene
No. of Wells 96
Format Strips of 8 Domed Caps
Product Type PCR Plate Cap Strip
View More Specs

Products 2
Catalog Number Mfr. No. Description Color No. per Case Price Quantity & Availability  
Catalog Number Mfr. No. Description Color No. per Case Price Quantity & Availability  
AB1810
View Documents
Thermo Scientific™
AB1810
Clear 25
Case of 25 for $91.00
Add to Cart
 
AB1810100
View Documents
Thermo Scientific™
AB1810/100
Clear 100
Case of 100 for $293.00
Add to Cart
 
Description

Description

Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

The Thermo Scientific VersiCap Mats are unique, with each strip of eight caps being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.

Highlights

  • Multiple cap strips can be applied at the same time when sealing qPCR plates, thereby reducing setup time
  • Remove the need to cut up adhesive seals
  • Suitable for use with all Thermo Scientific 96-well PCR plates as well as standard and low profile strip tubes
Applications
  • PCR
  • qPCR/Real-Time PCR

Autoclavable Resistance: Successfully Tested
DMSO (100%) Resistance: Successfully Tested
Ethanol Resistance: Successfully Tested
Gamma Irradiation Resistance: Successfully Tested
Hazardous: No
Isopropanol Resistance: Successfully Tested
Long Term Storage: Successfully Tested
PCR (including waterbath): Successfully Tested
Peelable: Successfully Tested
Pierceable: Not Recommended
qPCR:Ultra Clear caps: Successfully Tested
Domed or Flat caps: Not Recommended
Resealable: Successfully Tested
Sealing Temperature Range: −20°C to +120°C
Shelf Life: 60 Months
Shipping Condition: Ambient
Storage Condition: Ambient
UV Irradiation Resistance: Successfully Tested

  • PCR
  • qPCR/Real-Time PCR

Certifications

Certified free from human DNA, RNases, and DNases

Specifications

Specifications

Clear
96
PCR Plate Cap Strip
Yes
96-well Plate
Polypropylene
Strips of 8 Domed Caps
VersiPlate
-20°C to +120°C
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit