Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Verso 1-Step RT-PCR Kits

Get everything needed for RT-PCR in this complete set of reagents with a highly processive reverse transcriptase enzyme, available with regular or hot start Taq.

$242.50 - $1035.00

Specifications

For Use With (Application) 1-Step RT-PCR
Storage Requirements Store at -20°C. Avoid freeze/thaw cycles
View More Specs

Products 4
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Description Quantity Price Quantity & Availability  
AB1454LDA
View Documents
Thermo Scientific™
AB1454LDA
ReddyMix Master Mix with ThermoPrime Taq 80 x 25µL Rxns Each for $242.50
Add to cart
 
AB1455B
View Documents
Thermo Scientific™
AB1455B
Master Mix with Thermo-Start Taq (Hot Start) 400 x 25µL Rxns Each for $1,035.00
Add to cart
 
AB1455A
View Documents
Thermo Scientific™
AB1455A
Master Mix with Thermo-Start Taq (Hot Start) 80 x 25µL Rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
AB1454B
View Documents
Thermo Scientific™
AB1454B
Master Mix with ThermoPrime Taq 400 x 25µL Rxns This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Verso 1-Step RT-PCR system is a complete set of RT-PCR reagents that combines everything needed for RT-PCR. The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures. Verso also shows a better dynamic range than standard AMV or M-MuLV enzymes, with increases in template concentration giving rise to corresponding increases in product generated over a wider range of starting material concentrations.

The Verso system is available with ThermoPrime (standard PCR) or Thermo-Start (hot start) polymerase, the latter providing a more specific option that delivers higher yields and greatly reduces nonspecific primer extension before PCR. Thermo-Start Taq DNA Polymerase is a chemically-modified polymerase that remains inactive throughout the RT step, activated only by the high-temperature incubation (95°C for 15 minutes) prior to PCR.

An optional RT Enhancer is provided with kits and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment.

The Verso 1-step RT-PCR system is also available with a ReddyMix option, where the master mix contains a dye and precipitant that allow RT-PCR reactions to be loaded directly onto an electrophoresis gel. The red dye migrates between bromophenol blue and xylene cyanol at approximately 300 bp, depending on agarose concentration.

Highlights

  • Eliminate the need for DNase treatments -RT Enhancer (optional) prevents gDNA carry-over
  • Enzyme choice -Standard PCR or hot start options available
  • Available as ReddyMix formulation -eliminate post-PCR steps required for electrophoresis
  • RT and PCR steps performed in single sealed tube -convenience and reduced contamination risk
  • Verso enzyme is highly sensitive in a broad dynamic range -accurately detect RNA input from 1 pg to 1 μg
  • Wide working temperature range (42 to57°C) for success with GC-rich and other difficult templates
  • RNase Inhibitor included in RT enzyme mix
Specifications

Specifications

1-Step RT-PCR
Store at -20°C. Avoid freeze/thaw cycles

Purchase of this product includes an immunity from suit under patents specified in the product insert to use only the amount purchased for the purchaser's own internal research. No other patent rights are conveyed expressly, by implication, or by estoppel. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit