Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VERTEC™ LAC2000, Thermo Scientific™

Catalog Number AAH3112314
Click to view available options
Quantity:
5 g
25 g
100 g
96471-81-7
C14H32N2O4Ti
340.287
MFCD11113033
ILOYOQJSNBNOPE-UHFFFAOYSA-N
vertec? lac2000
90477800
1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol;titanium
CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O.[Ti]
96471-81-7
C14H32N2O4Ti
340.287
MFCD11113033
ILOYOQJSNBNOPE-UHFFFAOYSA-N
vertec? lac2000
90477800
1-[2-[bis(2-hydroxypropyl)amino]ethyl-(2-hydroxypropyl)amino]propan-2-ol;titanium
CC(CN(CCN(CC(C)O)CC(C)O)CC(C)O)O.[Ti]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Formula Weight 336.26
Chemical Name or Material VERTEC™ LAC2000
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit