Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VETRANAL™ 17-Hydroxyprogesterone, MilliporeSigma™ Supelco™

$64.10 - $99.00

Specifications

Product Line VETRANAL
Grade Analytical Standard
Linear Formula C21H30O3
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-5185
View Documents
MilliporeSigma Supelco
4633710MG
10 mg
Each for $64.10
Add to cart
 
11-101-5186
View Documents
MilliporeSigma Supelco
4633750MG
50 mg
Each for $99.00
Add to cart
 
Description

Description

Pharmaceutical (small molecule)
Specifications

Specifications

VETRANAL
Analytical Standard
C21H30O3
Neat
Safety and Handling

Safety and Handling

ShelfLife : Limited shelf life, expiry date on the label

RTECSNumber : TU5060000

SDS
Product Certifications

VETRANAL is a trademark of Sigma-Aldrich Chemie GmbH

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit