Learn More
VGAT Monoclonal Antibody (D9), FITC, Invitrogen™
Mouse Monoclonal Antibody
Supplier:  Thermo Scientific MA545925
Encompass
Catalog No. PIMA545925
Add to Cart
This item is not returnable. View return policy
Description
Description
VGAT Monoclonal antibody specifically detects VGAT in Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VGAT
Monoclonal
1 mg/mL
9.1mM sodium bicarbonate, 9.44mM PBS with 123mM ethanolamine, 581mM DMSO and no preservative; pH 7.4
O35458, O35633
SLC32A1
Synthetic peptide from the N-terminal region of Mouse VGAT protein
100 μg
Primary
Mouse, Rat
Liquid
ELISA, Immunohistochemistry, Western Blot
D9
FITC
SLC32A1
GABA and glycine transporter; GVAT; hVIAAT; mVGAT; mVIAAT; R75019; rGVAT; SLC32A1; solute carrier family 32 (GABA vesicular transporter), member 1; solute carrier family 32 member 1; vasicular inhibitory amino acid transporter 10D; vesicular GABA transporter; Vesicular inhibitory amino acid transporter; Vgat; Viaat
Mouse
Protein A
RUO
22348, 83612
4° C, store in dark
IgG2a
Recommendations
Recommendations