Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGLL2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP2843270.1ML

Catalog No. NB032255


Add to cart

Description

Description

VGLL2 Polyclonal specifically detects VGLL2 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

VGLL2
Unconjugated
Purified
Rabbit
Immunogen affinity purified
RUO
Primary
245806
Western Blot
PBS, 2% Sucrose
Protein VITO1, transcription cofactor vestigial-like 2, transcription cofactor vestigial-like protein 2, vestigial like 2 (Drosophila), vgl-2, VGL2, VITO1vestigial-like 2
The immunogen is a synthetic peptide corresponding to a region of Mouse VGLL2
0.1ml
Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit