Learn More
VGLUT1 Mouse anti-Human, Mouse, Rat, DyLight 755, Clone: S28-9, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259329IR
Catalog No. NBP259329IR
Add to cart
Description
Description
VGLUT1 Monoclonal antibody specifically detects VGLUT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
VGLUT1
S28-9
Western Blot, Immunohistochemistry
Purified
SLC17A7
Fusion protein amino acids 493-560 (cytoplasmic C-terminus) of rat VGlut1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 52kDa. No cross-reactivity against VGlut2.
Western Blot, Immunohistochemistry
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
BNPIbrain-specific Na-dependent inorganic phosphate cotransporter, Brain-specific Na(+)-dependent inorganic phosphate cotransporter, member 7, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), Solute carrier family 17 member 7, VGluT1, VGLUT1vesicular glutamate transporter 1
Mouse
IgG1
100 ul
Primary
57030
Human, Mouse, Rat