Learn More
VGluT1 Rabbit anti-Human,Mouse,Rat, KHL, Clone: 4O15, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB23744X25UL
 View more versions of this product
Catalog No. ZRB23744XMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
VGluT1 Monoclonal antibody specifically detects VGluT1 in Human,Mouse,Rat samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
VGluT1
4O15
KHL
Vesicular glutamate transporter 1;Brain-specific Na(+)-dependent inorganic phosphate cotransporter;Solute carrier family 17 member 7
Rabbit
IgG
Protein A Purified
Neuroscience
Primary
4X
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
Slc17a7, Bnpi, Vglut1
KLH-conjugated linear peptide corresponding to 19 amino acids from the C-terminal region of rat Vesicular glutamate transporter 1 (VGlut1).
∼53 kDa observed; 61.66 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human, Mouse, Rat