Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VGLUT2 Antibody (S29-29), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259330B
Description
VGLUT2 Monoclonal specifically detects VGLUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
VGLUT2 | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
SLC17A6 | |
Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2 | |
100 μL | |
57084 | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
S29-29 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, differentiation-associated Na-dependent inorganic phosphate cotransporter, DNPI, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2, VGLUT2 | |
Mouse | |
Protein G purified | |
Primary | |
Detects 60kDa. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction