Learn More
VGLUT2 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: S29-29, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259330C
Catalog No. NBP259330C
Add to cart
Description
Description
VGLUT2 Monoclonal antibody specifically detects VGLUT2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
VGLUT2
S29-29
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SLC17A6
Fusion protein amino acids 501-582 (cytoplasmic C-terminus) of rat VGlut2
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 60kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, differentiation-associated Na-dependent inorganic phosphate cotransporter, DNPI, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2, VGLUT2
Mouse
IgG1
100 ul
Primary
57084
Human, Mouse, Rat