Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VGLUT2 Rabbit anti-Mouse,Rat, KHL, Clone: 3C2, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen VGLUT2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB117025MI
View Documents
MilliporeSigma
ZRB117025UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB11704XMI
View Documents
MilliporeSigma
ZRB11704X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

VGLUT2 Monoclonal antibody specifically detects VGLUT2 in Mouse,Rat samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

VGLUT2
KHL
Rabbit
Vesicular glutamate transporter 2;Differentiation-associated BNPI;Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter;Solute carrier family 17 member 6
KLH-conjugated linear peptide corresponding to 18 amino acids from the C-terminal region of Rat Vesicular glutamate transporter 2 (VGLUT2).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Purified
Mouse, Rat
Slc17a6, Dnpi, Vglut2
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit