Learn More
VGluT3 Mouse anti-Human, Mouse, Rat, Biotin, Clone: S34-34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259331B
Catalog No. NBP259331B
Add to cart
Description
Description
VGluT3 Monoclonal antibody specifically detects VGluT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
VGluT3
S34-34
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SLC17A8
Fusion protein amino acids 546-588 (cytoplasmic C-terminus) of rat VGlut3
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 65kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8
Mouse
IgG1
100 ul
Primary
246213
Human, Mouse, Rat