Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
VGluT3 Mouse anti-Human, Mouse, Rat, DyLight 650, Clone: S34-34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259331C
Catalog No. NBP259331C
Description
Description
VGluT3 Monoclonal antibody specifically detects VGluT3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
VGluT3
S34-34
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SLC17A8
Fusion protein amino acids 546-588 (cytoplasmic C-terminus) of rat VGlut3
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 65kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 8
Mouse
IgG1
100 ul
Primary
246213
Human, Mouse, Rat