Learn More
Tocris Bioscience™ VH 298
High-affinity inhibitor of VHL
$351.48 - $1473.20
Specifications
Product Type | Inhibitor |
---|---|
Target Proteins | Ubiquitin Ligase (E3) |
Molecular Formula | C27H33N5O4S |
Formula Weight | 523.65g/mol |
Content And Storage | Store at -20°C |
Description
High-affinity inhibitor of E3 ubiquitin ligase VHL (Kd = 80-90 nM). Blocks interaction between VHL and HIF-α downstream of HIF-α hydroxylation, initiating hypoxic response. Results in time- and concentration-dependent accumulation of hydroxylated HIF-α, and upregulates mRNA and protein levels of HIF target genes, with similar transcriptional profile to hypoxia. Cell permeable.Negative control cis VH 298 also available.
Specifications
Inhibitor | |
C27H33N5O4S | |
Store at -20°C |
Ubiquitin Ligase (E3) | |
523.65g/mol | |
Ubiquitin Ligase (E3) Inhibitors |
For Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.