Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 165381AP

Catalog No. 50-173-1014


Add to cart

Description

Description

Von Hippel-Lindau syndrome (VHL) is a domitly inherited familial cancer syndrome predisposing to a variety of maligt and benign tumors. A germline mutation of this gene is the basis of familial inheritance of VHL syndrome. The protein encoded by this gene is a component of the protein complex that includes elongin B, elongin C, and cullin-2, and possesses ubiquitin ligase E3 activity. This protein is involved in the ubiquitination and degradation of hypoxia-inducible-factor (HIF), which is a transcription factor that plays a central role in the regulation of gene expression by oxygen. RNA polymerase II subunit POLR2G/RPB7 is also reported to be a target of this protein. Alternatively spliced transcript variants encoding distinct isoforms have been observed.
Specifications

Specifications

VHL
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P40337, P40338, Q64259
HRCA1, Protein G7, pVHL, RCA1, VHL, VHL1, Von Hippe-Lindau
Rabbit
IgG
150 μL
-20°C
Primary
22346, 24874, 7428
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
VHL
Liquid
VHL
VHL Fusion Protein Ag9843
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit