Learn More
Thermo Scientific™ Vial, Cuvette and Test Tube Holders for Orion™ AQ7100 and AQ8100 Spectrophotometers
Quickly adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels
Supplier: Thermo Scientific™ AQX1SQVH
Description
Easy to change & clean Orion™ AquaMate 7100 & 8100 Spectrophotometer sample holders are designed for maximum flexibility.
Easily adapt your Orion™ AquaMate 7100 & 8100 Spectrophotometer to work with multiple sample vessels
Magnetic retention makes these holders quick and easy to clean and change to adapt to your analytical needs.
- AQX1RNDVH: Vial / Test Tube Holder for vials and test tube from 12 to 25 mm OD for Orion™ AquaMate 7100 & 8100 Spectrophotometers
- AQX1SQVH: Standard 10 mm square single-cell holder for Orion™ AquaMate 7100 & 8100 Spectrophotometers
- AQX1LWLVH: Long path rectangular cell holder for 20-100 mm pathlength cuvettes for Orion™ AquaMate 7100 & 8100 Spectrophotometers
Specifications
10 mm square single-cell holder | |
Orion AquaMate 7100 & 8100 Spectrophotometers | |
1 square cell holder |
Single Cell Sample Holder | |
10 mm cuvette (1) | |
Square |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.