Learn More
Tocris Bioscience Vialinin A
Inhibitor of USP4 and USP5/IsoT
Manufacturer: Tocris Bioscience 4988/1
Description
Inhibitor of ubiquitin-specific peptidase 4 (USP4), USP5/isopeptidase T (IsoT) and UCH-L1 deubiquitinating enzyme (DUB) activity (IC50 values are 1.5, 5.9 and 22.3 μM, respectively). Shows no significant inhibition of UCH-L3, USP2, and USP8 activities. Decreases release of TNF-α (IC50 = 0.09 nM) from RBL-2H3 mast cells. Displays anti-cancer activity decreasing viability in human colonic carcinoma cells.This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Vialinin A | |
>98 | |
1mg |
Vialinin A | |
562.57 |