Learn More
Cole-Parmer Balmac Inc. 191-2 Vibration Transmitter, 0 to In./Sec  DFS Item
Manufacturer:  Cole-Parmer 9885315
FEATURES
  • Solid-state circuitry
  • Two-wire 4-20 mA signal format
  • Same wiring for both power and vibration signal
  • Economical for continuous monitoring of all machines
  • Connects in series with DC power supply and measurement devices
  • Works with single-loop monitors
  • Rugged NEMA 4 weatherproof case
APPLICATIONS
  • Fans, Motors, Pumps and Compressors
  • Controlling Vibrating Conveyors
  • General Purpose Machinery
  • Hazardous Environments
Catalog No. 50-627-385