Learn More
Vibrio Cholera Toxin Beta Subunit Mouse anti-Bacteria, Clone: 1C3, Abnova™
Mouse monoclonal antibody raised against Vibrio Cholera Toxin Beta Subnuit.
Manufacturer:  Abnova Corporation MAB12438
Catalog No. 89-935-083
Add to cart
Description
Description
Specifications
Specifications
Vibrio Cholera Toxin Beta Subunit
1C3
Mouse monoclonal antibody raised against Vibrio Cholera Toxin Beta Subnuit.
Lyophilized from PBS, pH 7.2 (0.09% sodium azide)
Mouse
IgG2b
1 mg
Store at 4°C on dry atmosphere. After reconstitution with deionized water, store at −20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA, Western Blot
Unconjugated
ELISA Western Blot The optimal working dilution should be determined by the end user.
Lyophilized
Native purified Vibrio Cholera Toxin Beta Subnuit.
Antigen Affinity Purification
RUO
Primary
Bacteria