Vidaro™ B-Fore™ Cleanroom Coveralls
Minimize particluate contamination and dissipates electrostatic charges
Manufacturer: Vidaro CorporationSupplier Diversity Partner C33587/L
 View more versions of this product
Catalog No. 19-999-826
  Check Availability
Add to Cart
    Specifications
    Material Polyester
    Color Light Blue
    Size Large
    Size (English) Chest 42 to 44 in.
    Size (Metric) Chest 106.6 to 111.7cm
    Closure Type Zipper
  • Description & Specifications
Related Products
Product Content Feedback