Vidaro™ B-Fore™ ESD Hoods
High-performance fabric
Manufacturer: Vidaro CorporationSupplier Diversity Partner H3158/M
 View more versions of this product
Catalog No. 19-999-881
  Check Availability
Add to Cart
Required Accessories : Two female snaps on each cheek allow the attachment of a facial (not included).
    Specifications
    Hood Material B-Fore (Polyester, Carbon)
    Quantity Each
    Includes Two snap on each cheek, Stainless Steel front-snap closure, snap at back of neck for adjustment
    Material B-Fore (Polyester, Carbon), Stainless Steel
    Size Medium
    Closure Type Snap front
    Color White
  • Description & Specifications
Related Products
Product Content Feedback