Learn More
Vidaro Polyester Taffeta Shoe Covers
Manufacturer:  Vidaro CorporationSupplier Diversity Partner ESC3111T/XL
 View more versions of this product
Catalog No. 19-130-0868D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Elastic
X-Large
Ideal for the microelectronics industry, contamination control, increase worker comfort, protect from static dissipative charges, airborne particles, lint
48cm
33.6cm
White
Polyester, CSM™ Rubber
19 in.
13.25 in.