Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vigabatrin, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

60643-86-9
C6H11NO2
129.159
PJDFLNIOAUIZSL-UHFFFAOYSA-N
4-Aminohexenoic acid
5665
CHEBI:63638

GABA-T inhibitor

Selective GABA-T (transaminase) inhibitor. Anticonvulsant.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

SMILES C=CC(CCC(=O)O)N
IUPAC Name 4-aminohex-5-enoic acid
Formula Weight 129.16
Quantity 10mg
Chemical Name or Material Vigabatrin
Grade >98%

Recommended Storage : Desiccate at +4°C