Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ 16 in. Foam Blade Squeegee

Remco™ Vikan™ 16 in. Foam Blade Squeegee is a floor squeegee idealfor drying surfaces after a sanitizing process.

$38.35 - $39.15

Specifications

Certifications/Compliance FDA Compliant
Description Thread type Euro Hygienic
Material Polypropylene, Foam Rubber
Quantity 1 Ea.
Size Large
View More Specs

Products 5
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
17-227-901
View Documents
Remco™
77524
Red
Each for $38.35
Add to Cart
 
17-227-903
View Documents
Remco™
77526
Yellow
Each for $38.35
Add to Cart
 
17-227-900
View Documents
Remco™
77522
Green
Each for $38.35
Add to Cart
 
19-830-222
View Documents
Remco™
77523
Blue
Each for $39.15
Add to Cart
 
17-227-902
View Documents
Remco™
77525
White
Each for $38.35
Add to Cart
 
Description

Description

  • Great on all types of floors
  • The soft rubber reaches into and cleans all spaces on the floor, such as grouting between tiles
  • The cartridge system blades can be removed for easy cleaning and replaced when worn out
  • Economical
  • May be used with any European-thread handle
  • Color-coded
Specifications

Specifications

FDA Compliant
Polypropylene, Foam Rubber
Large
Thread type Euro Hygienic
1 Ea.
3.5 in.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit