Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ 26 in. Basic Aluminum Handle

Remco™ Vikan™ 26 in. Basic Aluminum Handle is ideal for cleaning applications where a full-size handle isn't necessary.

$17.96

Specifications

Certifications/Compliance FDA Compliant
Description Thread type Euro Hygienic
Material Aluminum, Polypropylene
Type Handle
Sterility Non-sterile
View More Specs

Products 5
Catalog Number Mfr. No. Color Code Price Quantity & Availability  
Catalog Number Mfr. No. Color Code Price Quantity & Availability  
17-227-150
View Documents
Remco™
29816
Yellow
Each for $17.96
Add to cart
 
17-227-146
View Documents
Remco™
29812
Green
Each for $17.96
Add to cart
 
17-227-148
View Documents
Remco™
29814
Red This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-227-147
View Documents
Remco™
29813
Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
17-227-149
View Documents
Remco™
29815
White This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Ideal for applications where hygiene and cross-contamination is a concern
  • Economic
  • Can be used to clean benchtop surfaces, drains, and other tasks that don't require a full-size handle
  • Color-coded
Specifications

Specifications

FDA Compliant
Aluminum, Polypropylene
Non-sterile
1
Thread type Euro Hygienic
Handle
25.5 in.
1.25 in.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit