Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ 59 in. Aluminum Handle

Remco™ Vikan™ 59 in. Aluminum Handle is long enough to be comfortable to use for most employees, and light enough to be ergonomically friendly.

Manufacturer:  Remco™ 29372

 View more versions of this product

Catalog No. 17-227-094


Add to cart

Description

Description

  • Color-coded
  • For use with all brooms, squeegees, and scrapers
  • The handle with the attached item should reach to the chin of the user to prevent occupational back injuries
Specifications

Specifications

Green
Non-sterile
Handle
Aluminum, Polypropylene
1.25 in.
Yes
Thread type Euro Hygienic
FDA Compliant
59.25 in.
1
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit