Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Remco™ Vikan™ Long Handle Soft Brush

Designed to reach into parts of equipment that are hard to access.

Manufacturer:  Remco™ 41824

 View more versions of this product

Catalog No. 19-830-031


Add to cart

Description

Description

  • Polypropylene block with polyester bristles
  • Angled bristles with medium stiffness
Specifications

Specifications

121°C
Medium stiff
15.5 in.
allows to reach into parts of equipment that are hard to access
L x W x H: 15.5 x 2.75 x 2.8 in.
Polyester
Red
Long handle soft brush
Polypropylene
0.5 lb.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit