Learn More
Bel-Art™ SP Scienceware™ Vikem Vinyl Aprons
Provides protection to personnel in hospitals, schools, laboratories and industrial plants.
$40.80
Specifications
Color Clear
Construction Stitched cotton binding securing all edges
Material Vinyl
View More Specs
Products 1
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Length (Metric) Price Quantity & Availability  
22-261314
View Documents
Bel-Art™ SP Scienceware™
H246030001
91 cm Each for $40.80
Add to cart
Description
Description
  • Lightweight and comfortable
  • Made from 4mil thick vinyl
  • Stitched cotton binding securing all edges
  • Strong tie backs double reinforced with metal grommets through three layers of material at waist
  • Non-absorbent and chemically resistant
Specifications
Specifications
Clear
Stitched cotton binding securing all edges
Vinyl