Learn More
Freudenberg™ UltraSpeed Pro Breakaway Mop Frame
Developed to be used with UltraSpeed Pro System. Vileda Professional-FHP UltraSpeed Pro Breakaway Mop Frame is part of a comprehensive floor cleaning system that sets new standards in usability, versatility, efficiency and ergonomics.
Supplier:  Freudenberg™ 146963
Catalog No. 19-000-671
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Lightweight, durable, autoclavable
  • 40cm/16 in. breakaway can be used with UltraSpeed pads, standard pocket mops and flat sponge cleanroom mops
  • Includes a modern swivel joint with integrated ball catch, making it easy to clean vertical surfaces
  • Easy, fast rinsing and wringing of mops without removal from frame
  • Attaches to Vileda Extendable Handle
Specifications
Specifications
16 in.
10/Cs.
Ideal for Use in Laboratory, LAR, Production and ISO 7 Enivronments
Surface Cleaning in LAR, Production, and Lab Environments
Product Suggestions
Product Suggestions