Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Villin 1, Mouse anti-Human, Alexa Fluor 647, Clone: rVIL1/1325, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279943AF647
Description
Villin 1 Monoclonal antibody specifically detects Villin 1 in Human Samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Villin 1 | |
rVIL1/1325 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
VIL1 | |
A recombinant fragment (around aa179-311) of human Villin 1 protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
This antibody recognizes a protein of 95kDa, which is identified as Villin 1. It is a major constituent in the microvilli, which compose the brush border of epithelial cells forming absorptive surfaces of the intestinal and renal proximal tubular epithelia. Anti-Villin 1 labels the brush border area in the gastrointestinal mucosal epithelium and urogenital tract. Among neoplasms, villin is predominantly expressed in tumors of colorectal origin. This antibody is useful in identifying malignant cells from primary and metastatic colorectal carcinomas; it also labels Merkel cells of the skin. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
D2S1471, VIL, villin 1, villin-1 | |
Mouse | |
IgG1 Kappa | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers | |
Primary | |
7429 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title