Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villin 1 Rabbit anti-Human, Clone: VIL1/2310R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Villin 1
Clone VIL1/2310R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121841
View Documents
Novus Biologicals
NBP30718410UG
100 μg
Each for $406.52
Add to cart
 
NB121842
View Documents
Novus Biologicals
NBP30718420UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

Villin 1 Monoclonal antibody specifically detects Villin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Villin 1
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
D2S1471, VIL, villin 1, villin-1
A recombinant fragment (around aa179-311) of human Villin 1 protein (exact sequence is proprietary)
Monoclonal
Protein A or G purified
VIL1/2310R
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
7429
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit