Learn More
Villin Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS3810R
Catalog No. 50-198-5673
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Villin is part of cytoskeleton associated with the microvillar actin core bundle of gastrointestinal and renal brush border. It can interact with actin in a Ca2+ and phosphoinositide-regulated manner. Villin is a very specific marker for adenocarcinomas of the gastrointestinal tract and the pancreas. It is also expressed in some Merkel cell tumor and adenocarcinoma of the lung, prostate, ovarian and kidney.
Specifications
Specifications
Villin
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
VIL1
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
7429
VIL1
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P09327
D2S1471; OTTHUMP00000207159; VIL; VIL1; villin 1; Villin-1
KLH conjugated synthetic peptide derived from human Villin.
Protein A
RUO
Antibody
Polyclonal
Human