Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Villin1 Polyclonal Antibody, Huabio
Rabbit Polyclonal Antibody
Supplier: HUABIO RESEARCH INC ER190131


Description
Villin1 Polyclonal antibody specifically detects Villin1 in Human, Mouse samples. It is validated for Flow Cytometry, Western BlotSpecifications
Villin1 | |
Polyclonal | |
Unconjugated | |
VIL1 | |
D2S1471; OTTHUMP00000207159; Vil; Vil1; villin 1; villin-1 | |
Synthetic peptide within human Villin aa 800-827. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P09327, Q62468 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22349, 7429 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction