Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VIM Mouse anti-Human, Clone: LN-6, Abnova™

Mouse monoclonal antibody raised against native human VIM.

Manufacturer:  Abnova Corporation MAB13451

Catalog No. 89-935-092


Add to cart

Description

Description

This gene encodes a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein encoded by this gene is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a dominant, pulverulent cataract

Specifications

Specifications

VIM
LN-6
Mouse monoclonal antibody raised against native human VIM.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
FLJ36605
Mouse
IgM
100 ug
Store at 4°C.
Monoclonal
This monoclonal antibody reacts with a non-hematopoietic epitope of VIM and shows no cross-reaction with other closely related intermediate filament proteins such as desmin, keratin, neurofilament, and glial fibrillary acid protein.
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
VIM
Human thymic nuclear extract.
PEG precipitation
RUO
Primary
7431
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit