Learn More
VIM Mouse anti-Human, Clone: OTI1G6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801360
Catalog No. 50-166-5565
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.
Specifications
Specifications
VIM
Monoclonal
Unconjugated
VIM
Lyophilized
VIM
Full length human recombit protein of human VIM produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1G6
PBS with 8% trehalose and no preservative; pH 7.3
P08670
vimentin
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
7431