Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Vimentin Ab-2, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Vimentin Ab-2, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ MS129R7

 View more versions of this product

Catalog No. MS129R7

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Vimentin Ab-2, Mouse Monoclonal Antibody.

Vimentin is the main intermediate filament protein in mesenchymal cells and is therefore of value in the differential diagnosis of undifferentiated neoplasms.

Host Species: Mouse

Clone: V9

Isotype: IgG1/κ

Species Reactivity: Rat, Horse, Chicken, Cow, Cat, Dog, Human and Pig. Others not known.

Epitope: Not determined

Immunogen: Purified vimentin from pig eye lens

Molecular Weight: 57-60kDa

Positive Control: Sarcomas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Vimentin Ab-2
Monoclonal
Unconjugated
Prediluted
Purified vimentin from pig eye lens
7mL
Cytoskeletal Protein
Primary
Bovine, Canine, Chicken, Equine, Feline, Gerbil, Hamster, Human, Murine, Non-human Primate, Porcine, Rabbit, Rat
Immunohistochemistry (Paraffin)
V9
Ready-to-use for IHC (UVLP)
Murine
IgG2a kappa
IVD
Primary
Monoclonal
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit