Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Vimentin

$1787.72

Specifications

Molecular Weight 57
Name Vimentin
Packaging Poly Micro Tube
View More Specs

Products 1
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP2545100
View Documents
100μg
Each for $1,787.72
Case of 5 EA for $7,181.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

Specifications

Specifications

57
Vimentin
Poly Micro Tube
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit