Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Mouse anti-Human, Mouse, Rat, DyLight™ 550, Clone: 2D1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192687R

Catalog No. NBP192687R


Add to cart

Description

Description

Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Vimentin
2D1
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
VIM
Recombinant human Vimentin expressed in and purified from E. coli. [UniProtNo. P08670]
Protein G purified
Biologically Active Proteins, Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells
Primary
7431.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
DyLight™ 550
50 mM sodium borate with 0.05% sodium azide
FLJ36605, vimentin
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit