Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Mouse anti-Human, Mouse, Rat, Porcine, Canine, Chicken, Goat, Hamster, Primate, Xenopus, Zebrafish, PE-Cyanine7, Clone: RV202, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP197672PECY7
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Mouse, Rat, Porcine, Canine, Chicken, Goat, Hamster, Primate, Xenopus, Zebrafish samples. It is validated for Flow Cytometry.Specifications
Vimentin | |
RV202 | |
Flow Cytometry | |
Conjugated, Purified | |
VIM | |
Clone RV202 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a cytoskeletal vimentin extract of calf lens. | |
Protein A or G purified | |
Biologically Active Proteins, Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells | |
Primary | |
7431.0 | |
Canine, Chicken, Goat, Hamster, Human, Mouse, Porcine, Primate, Rat, Xenopus, Zebrafish |
Flow Cytometry | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
FLJ36605, vimentin | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. Do not freeze. | |
Monoclonal | |
RV202 reacts exclusively with vimentin, which is expressed in mesenchymal cells and mesenchymal derived tumors e.g. lymphoma, sarcoma and melanoma. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title