Learn More
Vimentin Mouse anti-Human, Rat, DyLight 488, Clone: 2A52, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192688G
Catalog No. NBP192688G
Add to cart
Description
Description
Vimentin Monoclonal antibody specifically detects Vimentin in Human, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown.
Specifications
Specifications
Vimentin
2A52
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
VIM
Full length recombinant human vimentin protein expressed in and purified from E. coli.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Rat
Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
FLJ36605, vimentin
Mouse
IgG1
0.1 ml
Biologically Active Proteins, Cancer, Cellular Markers, Cytoskeleton Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Markers, Stem Cells
Primary
7431