Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinculin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10
Specifications
Antigen | Vinculin |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 0.5-2 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunofluorescence 20 μg/mL, Immunocytochemistry 5 μg/mL |
Applications | Western Blot, ELISA, Immunofluorescence, Immunofluorescence, Immunocytochemistry |
Classification | Polyclonal |
Description
Vinculin Polyclonal antibody specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, ImmunocytochemistrySpecifications
Vinculin | |
Western Blot 0.5-2 μg/mL, ELISA, Immunocytochemistry/ Immunofluorescence 5-20 μg/mL, Immunofluorescence 20 μg/mL, Immunocytochemistry 5 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
Antibody was raised against a peptide corresponding to 23 amino acids near the center of human Vinculin. < Amino Acid Sequence: VREAFQPQEPDFPPPPPDLEQLR | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot, ELISA, Immunofluorescence, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Cell Biology, Cellular Markers | |
PBS | |
7414 | |
IgG | |
Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title